Novus Biologicals
Manufacturer Code:NB10077303
Catalog # NB10077303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 900 and 950 of human WD repeat domain 62 using the numbering given in entry NP_775907.4 (GeneID 284403). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C19orf14MGC166976 chromosome 19 open reading frame 14 DKFZP434J046 DKFZp686G1024 EC 2.7.11.1 EC 2.7.7.38 FLJ33298 WD repeat domain 62 WD repeat-containing protein 62; microcephaly, primary autosomal recessive 2; truncated WDR62; WD repeat-containing protein 62
Gene Aliases: C19orf14; MCPH2; WDR62
UniProt ID: (Human) O43379
Entrez Gene ID: (Human) 284403
Molecular Function:
RNA binding protein
enzyme modulator
esterase
hydrolase
kinase inhibitor
kinase modulator
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.