Invitrogen
Manufacturer Code:PA530833
Catalog # PIPA530833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 240 of Human WDR61 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: meiotic recombination protein REC14; Meiotic recombination REC14 protein homolog; REC14 SKI8 WD repeat-containing protein 61 meiotic recombination REC14 protein homolog recombination protein REC14; recombination protein REC14; Ski8; SKI8 homolog; WD repeat-containing protein 61; WD repeat-containing protein 61, N-terminally processed
Gene Aliases: 2700038L12Rik; 2810418I05Rik; REC14; SKI8; WDR61
UniProt ID: (Human) Q9GZS3, (Mouse) Q9ERF3
Entrez Gene ID: (Human) 80349, (Mouse) 66317
Molecular Function: RNA binding protein enzyme modulator esterase hydrolase kinase inhibitor kinase modulator mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.