Novus Biologicals
Manufacturer Code:NBP190438
Catalog # NBP190438
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ASLDESGVLNVWVVVELPKADIAGSISDLGLMPGGRVKLVHSALIQLGDSLSHKGNEFWGTTQTLNVKFLPSDPNHFIIGTDMGLISHGTRQDLRVAPKLFKPQQHGIRPVKVN |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cytoplasmic dynein 2 intermediate chain 1; Dynein 2 intermediate chain 1; FLJ10300 FLJ23575 WD repeat domain 60 WD repeat-containing protein 60; testicular secretory protein Li 66; WD repeat-containing protein 60
Gene Aliases: DYNC2I1; FAP163; SRPS6; SRTD8; WDR60
UniProt ID: (Human) Q8WVS4
Entrez Gene ID: (Human) 55112
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.