Abnova
Manufacturer Code:H00055112K
Catalog # 89-207-590
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human WDR60 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human WDR60 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Cytoplasmic dynein 2 intermediate chain 1; Dynein 2 intermediate chain 1; FLJ10300 FLJ23575; testicular secretory protein Li 66; WD repeat-containing protein 60
Gene Aliases: DYNC2I1; FAP163; SRPS6; SRTD8; WDR60
UniProt ID: (Human) Q8WVS4
Entrez Gene ID: (Human) 55112
Molecular Function:
cytoskeletal protein
microtubule family cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.