Invitrogen
Manufacturer Code:PA541984
Catalog # PIPA541984
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human WDR6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: mWDR6; WD repeat domain 6; WD repeat domain 6 protein; WD repeat protein 6; WD repeat-containing protein 6
Gene Aliases: mWDR6; WDR6
UniProt ID: (Bovine) A7Z052, (Human) Q9NNW5, (Rat) Q5XFW6, (Mouse) Q99ME2
Entrez Gene ID: (Horse) 100063394, (Bovine) 526884, (Dog) 608948, (Rabbit) 100354111, (Human) 11180, (Rat) 301007, (Mouse) 83669, (Guinea pig) 100727147
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.