Invitrogen
Manufacturer Code:MA515664
Catalog # PIMA515664
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 7B11 |
Immunogen | Purified recombit fragment of human WDR5 expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BMP2-induced 3-kb gene protein; cilia and flagella associated protein 89; SWD3 BIG-3 WDR5; SWD3, Set1c WD40 repeat protein, homolog; WD repeat-containing protein 5
Gene Aliases: BIG-3; BIG3; CFAP89; SWD3; WDR5
UniProt ID: (Human) P61964
Entrez Gene ID: (Human) 11091
Molecular Function:
RNA binding protein
enzyme modulator
esterase
hydrolase
kinase inhibitor
kinase modulator
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.