Invitrogen
Manufacturer Code:PA5102553
Catalog # PIPA5102553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human WDR5 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2410008O07Rik AA408785 AA960360 Big BIG3 BIG-3 BMP2-induced 3-kb gene protein CFAP89 cilia and flagella associated protein 89 SWD3 SWD3 Set1c WD40 repeat protein homolog WD repeat domain 5 WD repeat-containing protein 5 WD repeat-containing protein BIG-3 Wdr5; BMP2-induced 3-kb gene protein; cilia and flagella associated protein 89; SWD3, Set1c WD40 repeat protein, homolog; WD repeat-containing protein 5; WD repeat-containing protein BIG-3
Gene Aliases: 2410008O07Rik; AA408785; AA960360; Big; BIG-3; BIG3; CFAP89; SWD3; WDR5
UniProt ID: (Human) P61964, (Mouse) P61965, (Rat) Q498M4
Entrez Gene ID: (Human) 11091, (Mouse) 140858, (Rat) 362093
Molecular Function: RNA binding protein enzyme modulator esterase hydrolase kinase inhibitor kinase modulator mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.