Abnova
Manufacturer Code:PAB14068
Catalog # 89-116-908
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A mixture of synthetic peptides (conjugated with KLH) that corresponding to internal region of mouse Wdr5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Wdr5.
For Research Use Only
Protein Aliases: 2410008O07Rik; AA408785; AA960360; Big; Big-3; BMP2-induced 3-kb gene protein; WD repeat-containing protein 5; WD repeat-containing protein BIG-3
Gene Aliases: 2410008O07Rik; AA408785; AA960360; Big; Big-3; Big3; Wdr5
UniProt ID: (Mouse) P61965
Entrez Gene ID: (Mouse) 140858
Molecular Function:
RNA binding protein
enzyme modulator
esterase
hydrolase
kinase inhibitor
kinase modulator
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.