OriGene
Manufacturer Code:TA890061
Catalog # 50-168-4774
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human WDR48 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p80; P80 SPG60 UAF1; testicular tissue protein Li 224; USP1 associated factor 1; USP1-associated factor 1; WD repeat endosomal protein; WD repeat-containing protein 48
Gene Aliases: 8430408H12Rik; KIAA1449; mKIAA1449; P80; RGD1309702; SPG60; UAF1; WDR48
UniProt ID: (Human) B4DY84, (Mouse) Q8BH57
Entrez Gene ID: (Human) 57599, (Rat) 363164, (Mouse) 67561
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.