Novus Biologicals
Manufacturer Code:NBP304699100UL
Catalog # NB093376
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-360 of human WDR45 (NP_001025067.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: JM5 WD repeat domain 45 WD repeat domain phosphoinositide-interacting protein 4 WD repeat-containing protein 45 WD45 repeat protein interacting with phosphoinositides 4 WDRX1 WDRXI4 WIPI-4 X-linked 1; neurodegeneration with brain iron accumulation 4; neurodegeneration with brain iron accumulation 5; WD repeat domain phosphoinositide-interacting protein 4; WD repeat domain, X-linked 1; WD repeat-containing protein 45; WD45 repeat protein interacting with phosphoinositides 4; WIPI-4
Gene Aliases: JM5; NBIA4; NBIA5; WDR45; WDRX1; WDRXI4; WIPI-4; WIPI4
UniProt ID: (Human) Q9Y484
Entrez Gene ID: (Human) 11152
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.