Novus Biologicals
Manufacturer Code:NBP186901
Catalog # NBP186901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:AGSVGLALCGQTLVVRGGSRFLATSIASSDDDSLFIYDCSAAEKKSQENKGEDAPLDQGSGAILASTFSNSGSYFALTDDSKRLILFRTKPWQCLSVRTVAR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hWH; Protein Wuho homolog; TRM82 tRNA methyltransferase 82 homolog; tRNA (guanine-N(7)-)-methyltransferase non-catalytic subunit WDR4; tRNA (guanine-N(7)-)-methyltransferase subunit WDR4; tRNA (guanine-N(7)-)-methyltransferase subunit WDR4 WD repeat domain 4 10TRM82 WD repeat-containing protein 4; WD repeat-containing protein 4; WDR4
Gene Aliases: TRM82; TRMT82; WDR4
UniProt ID: (Human) P57081
Entrez Gene ID: (Human) 10785
Molecular Function:
RNA binding protein
mRNA processing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.