Invitrogen
Manufacturer Code:PA596650
Catalog # PIPA596650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-339 of human CIAO1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CIAO1; ciao1 {ECO:0000255|HAMAP-Rule:MF_03037}; cytosolic iron-sulfur protein assembly 1; cytosolic iron-sulfur protein assembly 1 homolog; Probable cytosolic iron-sulfur protein assembly protein CIAO1; probable cytosolic iron-sulfur protein assembly protein CIAO1 {ECO:0000255|HAMAP-Rule:MF_03037}; WD repeat domain 39; WD repeat-containing protein 39; WD repeat-containing protein 39 {ECO:0000255|HAMAP-Rule:MF_03037}; WD40 protein Ciao1
Gene Aliases: AW210570; CIA1; CIAO1; WDR39
UniProt ID: (Human) O76071, (Rat) Q5M7T1, (Mouse) Q99KN2
Entrez Gene ID: (Human) 9391, (Rat) 29231, (Mouse) 26371
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.