Abnova
Manufacturer Code:H00134430A01
Catalog # 89-107-616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 853 a.a. &bsim 951 a.a) partial recombinant protein with GST tag.,WDR36 (NP_644810 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant WDR36.
For Research Use Only
Protein Aliases: DKFZp686I1650; GLC1G; T-cell activation WD repeat protein; T-cell activation WD repeat-containing protein; TA-WDRP; TAWDRP; UTP21; WD repeat-containing protein 36
Gene Aliases: GLC1G; TA-WDRP; TAWDRP; UTP21; WDR36
UniProt ID: (Human) Q8NI36
Entrez Gene ID: (Human) 134430
Molecular Function:
RNA binding protein
mRNA processing factor
mRNA splicing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.