Invitrogen
Manufacturer Code:PA560629
Catalog # PIPA560629
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human WDR35 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Intraflagellar transport protein 121 homolog; Naofen; WD repeat-containing protein 35; WDR35 WD repeat-containing protein 35 CED2
Gene Aliases: 4930459M12Rik; 4931430C06; CED2; IFT121; IFTA1; KIAA1336; mKIAA1336; RGD1564116; SRTD7; WDR35
UniProt ID: (Human) Q9P2L0, (Mouse) Q8BND3, (Rat) A6N6J5
Entrez Gene ID: (Human) 57539, (Mouse) 74682, (Rat) 503018
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.