Novus Biologicals
Manufacturer Code:NBP160030
Catalog # NBP160030
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to WDR33(WD repeat domain 33) The peptide sequence was selected from the middle region of WDR33. Peptide sequence TKFVRTSTNKVKCPVFVVRWTPEGRRLVTGASSGEFTLWNGLTFNFETIL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: NET14 WD repeat domain 33 WD repeat-containing protein 33 WD repeat-containing protein WDC146 WDC146FLJ11294; pre-mRNA 3' end processing protein WDR33; WD repeat-containing protein 33; WD repeat-containing protein of 146 kDa; WD repeat-containing protein WDC146
Gene Aliases: NET14; WDC146; WDR33
UniProt ID: (Human) Q9C0J8
Entrez Gene ID: (Human) 55339
Molecular Function: RNA binding protein mRNA processing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.