Invitrogen
Manufacturer Code:PA524133
Catalog # PIPA524133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 113-142 amino acids from the N-terminal region of human WDR32 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DDB1- and CUL4-associated factor 10; WD repeat domain 32; WD repeat-containing protein 32; WDR32 DDB1- and CUL4-associated factor 10 WD repeat domain 32 WD repeat-containing protein 32
Gene Aliases: DCAF10; WDR32
UniProt ID: (Human) Q5QP82
Entrez Gene ID: (Human) 79269
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.