Novus Biologicals
Manufacturer Code:NBP29427020UL
Catalog # NB023096
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 362-661 of human WDR26 (NP_079436.4). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CUL4- and DDB1-associated WDR protein 2; GID complex subunit 7 homolog; MIP2 myocardial ischemic preconditioning up-regulated protein 2 WD repeat domain 26 WD repeat-containing protein 26; Myocardial ischemic preconditioning up-regulated protein 2; myocardial ischemic preconditioning upregulated protein 2; WD repeat-containing protein 26
Gene Aliases: CDW2; GID7; MIP2; PRO0852; WDR26
UniProt ID: (Human) Q9H7D7
Entrez Gene ID: (Human) 80232
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.