Novus Biologicals
Manufacturer Code:NBP17074320UL
Catalog # NBP17074320
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to WDR21B(WD repeat domain 21B) The peptide sequence was selected from the middle region of WDR21B. Peptide sequence HEEEGIVVAVGQDCYTRIWSLHDAHLLRTIPSPYSASEDDIPSVAFASRL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DDB1 and CUL4 associated factor 4-like 1; DDB1- and CUL4-associated factor 4-like protein 1; WD repeat domain 21B; WD repeat-containing protein 21B
Gene Aliases: DCAF4L1; WDR21B
UniProt ID: (Human) Q3SXM0
Entrez Gene ID: (Human) 285429
Molecular Function: RNA binding protein enzyme modulator esterase hydrolase kinase inhibitor kinase modulator mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.