Proteintech
Manufacturer Code:136471AP150UL
Catalog # 50-172-8905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | WDR19/IFT144 Fusion Protein Ag4599 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: IFT144 KIAA1638 ORF26 PWDMP WD repeat domain 19 WDR19; Intraflagellar transport 144 homolog; WD repeat membrane protein PWDMP; WD repeat-containing protein 19
Gene Aliases: ATD5; C330027H04Rik; CED4; D330023L08Rik; DYF-2; DYF2; IFT144; KIAA1638; mKIAA1638; NPHP13; ORF26; Oseg6; PWDMP; RGD1306997; SRTD5; WDR19
UniProt ID: (Human) Q8NEZ3, (Mouse) Q3UGF1
Entrez Gene ID: (Human) 57728, (Rat) 305349, (Mouse) 213081
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.