Novus Biologicals
Manufacturer Code:NBP184033
Catalog # NBP184033
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:TGADYIVKIFDRHGQKRSEINLPGNCVAMDWDKDGDVLAVIAEKSSCIYLWDANTNKTSQLDNGMRDQMSFLLWSKVGSFLAVGTVKG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DYF-2 FLJ23127 IFT144 intraflagellar transport 144 homolog KIAA1638Pwdmp ORF26 Oseg6 PWDMP WD repeat domain 19 WD repeat membrane protein PWDMP WD repeat-containing protein 19; Intraflagellar transport 144 homolog; WD repeat membrane protein PWDMP; WD repeat-containing protein 19
Gene Aliases: ATD5; CED4; DYF-2; IFT144; KIAA1638; NPHP13; ORF26; Oseg6; PWDMP; SRTD5; WDR19
UniProt ID: (Human) Q8NEZ3
Entrez Gene ID: (Human) 57728
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.