Invitrogen
Manufacturer Code:PA534377
Catalog # PIPA534377
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid synthetic peptide near the carboxy terminus of human WDR18 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Involved in Processing ITS2 3 homolog; WD repeat-containing protein 18; WDR18 WD repeat domain 18 WD repeat-containing protein 18
Gene Aliases: 2310012I10Rik; AU044733; AW122032; Ipi3; R32184_1; WDR18
UniProt ID: (Human) Q9BV38, (Rat) Q499N3, (Mouse) Q4VBE8
Entrez Gene ID: (Human) 57418, (Rat) 314617, (Mouse) 216156
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.