Novus Biologicals
Manufacturer Code:NBP220889
Catalog # NBP220889
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human WDR16. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cilia- and flagella-associated protein 52; WD repeat domain 16; WD repeat domain 16 WD repeat-containing protein 16 WD40-repeat protein upregulated in HCC WD40-repeat protein up-regulated in HCC WDRPUHFLJ37528; WD repeat-containing protein 16; WD40-repeat protein up-regulated in HCC; WD40-repeat protein upregulated in HCC
Gene Aliases: CFAP52; WDR16; WDRPUH
UniProt ID: (Human) B2RDU7
Entrez Gene ID: (Human) 146845
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.