Proteintech
Manufacturer Code:201221AP150UL
Catalog # 50-173-2713
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | WDR13 Fusion Protein Ag13793 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: memory-related protein; MG21 WD repeat domain 13 WDR13; WD repeat-containing protein 13
Gene Aliases: 1700060B08Rik; 5730411P10Rik; DXHXS7467e; MG21; mMg21; RGD1560982; W51679; WDR13
UniProt ID: (Human) Q06DX1, (Mouse) Q91V09
Entrez Gene ID: (Human) 64743, (Rat) 317370, (Mouse) 73447
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.