Invitrogen
Manufacturer Code:PA557635
Catalog # PIPA557635
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human WDR12 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: nuclear protein Ytm1; Ribosome biogenesis protein WDR12; WD repeat-containing protein 12; WDR12; YTM1 WD repeat-containing protein 12 ribosome biogenesis protein WDR12
Gene Aliases: 4933402C23Rik; MNCb-5414; WDR12; YTM1; Ytm1p
UniProt ID: (Human) Q9GZL7, (Mouse) Q9JJA4
Entrez Gene ID: (Human) 55759, (Mouse) 57750
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.