Novus Biologicals
Manufacturer Code:NBP294831100UL
Catalog # NB024172
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human WDR12 (NP_060726.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ10881 FLJ12719 FLJ12720 ribosome biogenesis protein WDR12 WD repeat domain 12 WD repeat-containing protein 12 YTM1; Ribosome biogenesis protein WDR12; WD repeat-containing protein 12; WDR12
Gene Aliases: WDR12; YTM1
UniProt ID: (Human) Q9GZL7
Entrez Gene ID: (Human) 55759
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.