Novus Biologicals
Manufacturer Code:NBP298001AF350
Catalog # NB050769
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human WDR1 fragment. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Actin-interacting protein 1; Actin-interacting protein 1 AIP1 NORI-1 WD repeat domain 1 WD repeat-containing protein 1; AIP1; epididymis secretory protein Li 52; NORI-1; WD repeat-containing protein 1
Gene Aliases: AIP1; HEL-S-52; NORI-1; WDR1
UniProt ID: (Human) O75083
Entrez Gene ID: (Human) 9948
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.