Proteintech
Manufacturer Code:175581AP150UL
Catalog # 50-173-1643
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | WDFY4 Fusion Protein Ag11720 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C10orf64 KIAA1607 WDFY family member 4 WDFY4; WD repeat- and FYVE domain-containing protein 4; WDFY family member 4
Gene Aliases: C10orf64; Gm18810; KIAA1607; RGD1564142; WDFY4
UniProt ID: (Human) Q6ZS81, (Mouse) E9Q2M9
Entrez Gene ID: (Human) 57705, (Rat) 498582, (Mouse) 545030
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.