Novus Biologicals
Manufacturer Code:NBP103332AF647
Catalog # NB103332AFX
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen recognized by this antibody maps to a region between residue 500 and 550 of human WD repeat and FYVE domain containing 3 using the numbering given in entry NP_055806.2 (GeneID 23001). |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alfy; ALFY Autophagy-linked FYVE protein KIAA0993MGC16461 WD repeat and FYVE domain containing 3 WD repeat and FYVE domain-containing protein 3 ZFYVE25; Autophagy-linked FYVE protein; WD repeat and FYVE domain-containing protein 3
Gene Aliases: ALFY; KIAA0993; WDFY3; ZFYVE25
UniProt ID: (Human) Q8IZQ1
Entrez Gene ID: (Human) 23001
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.