Proteintech
Manufacturer Code:550091AP150UL
Catalog # 50-173-5927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alfy; ALFY Autophagy linked FYVE protein KIAA0993 WDFY3 ZFYVE25; Autophagy-linked FYVE protein; Beach domain, WD repeat and FYVE domain-containing protein 1; BWF1; galactosyltransferase 3 beta 1, 4; WD repeat and FYVE domain-containing protein 3
Gene Aliases: 2610509D04Rik; ALFY; AW319683; B930017C24; Bchs; BWF1; D5Ertd66e; Ggtb3; KIAA0993; mKIAA0993; WDFY3; ZFYVE25
UniProt ID: (Human) Q8IZQ1, (Mouse) Q6VNB8
Entrez Gene ID: (Human) 23001, (Mouse) 72145
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.