Novus Biologicals
Manufacturer Code:NBP29420120UL
Catalog # NB022960
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human WDFY2 (NP_443182.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Prof; Propeller-FYVE protein; propeller-FYVE protein RP11-147H23.1 WD repeat and FYVE domain containing 2 WD repeat and FYVE domain-containing protein 2 WD40 and FYVE domain containing 2 WD40- and FYVE domain-containing protein 2 WDF2 ZFYVE22PROF Zinc finger FYVE domain-containing protein 22; WD repeat and FYVE domain-containing protein 2; WD40 and FYVE domain containing 2; WD40- and FYVE domain-containing protein 2; Zinc finger FYVE domain-containing protein 22
Gene Aliases: PROF; WDF2; WDFY2; ZFYVE22
UniProt ID: (Human) Q96P53
Entrez Gene ID: (Human) 115825
Molecular Function:
membrane traffic protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.