Novus Biologicals
Manufacturer Code:NBP304348
Catalog # NB094972
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human WBSCR17 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp434I2216 EC 2.4.1.41 GalNAc-T-like protein 3 GALNT16 GALNT20 GALNTL3DKFZp761D2324 Polypeptide GalNAc transferase-like protein 3 polypeptide N-acetylgalactosaminyltransferase WBSCR17 pp-GalNAc-transferase 20 pp-GaNTase-like protein 3 Protein-UDP acetylgalactosaminyltransferase-like protein 3 putative polypeptide N-acetylgalactosaminyltransferase-like protein 3 UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like 3 UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 3 UDP-N-acetyl-alpha-D-galactosamine:polypeptideN-acetylgalactosaminyltransferase 16 Williams-Beuren syndrome chromosomal region 17 protein Williams-Beuren syndrome chromosome region 17; GalNAc-T-like protein 3; polypeptide GalNAc transferase 3; Polypeptide GalNAc transferase-like protein 3; Polypeptide N-acetylgalactosaminyltransferase 17; polypeptide N-acetylgalactosaminyltransferase WBSCR17; polypeptide N-acetylgalactosaminyltransferase-like 3; pp-GalNAc-transferase 20; pp-GaNTase-like protein 3; Protein-UDP acetylgalactosaminyltransferase-like protein 3; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like 3; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 3; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 16; Williams-Beuren syndrome chromosomal region 17 protein
Gene Aliases: GalNAc-T5L; GALNACT17; GALNT16; GALNT17; GALNT20; GALNTL3; WBSCR17
UniProt ID: (Human) Q6IS24
Entrez Gene ID: (Human) 64409
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.