Invitrogen
Manufacturer Code:PA558612
Catalog # PIPA558612
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human WBP4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: domain-containing binding protein 4; formin binding protein 21; Formin-binding protein 21; WBP-4; WW domain binding protein 4 (formin binding protein 21); WW domain binding protein 4 (formin binding protein 21) FBP21 MGC117310; WW domain-binding protein 4; WW domain-containing binding protein 4; WW domain-containing-binding protein 4
Gene Aliases: AW545037; BB101031; FBP21; FNBP21; WBP4
UniProt ID: (Human) O75554, (Mouse) Q61048, (Rat) Q5HZF2
Entrez Gene ID: (Human) 11193, (Mouse) 22380, (Rat) 114765
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.