Invitrogen
Manufacturer Code:PA5110577
Catalog # PIPA5110577
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-376 of human WBP4 (NP_009118.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW545037 BB101031 BOS_12322 FBP21 FNBP21 Formin-binding protein 21 MGC117310 Wbp4 WBP-4 WW domain-binding protein 4 WW domain-containing-binding protein 4 zgc:112384; domain-containing binding protein 4; formin binding protein 21; Formin-binding protein 21; WBP-4; WW domain binding protein 4 (formin binding protein 21); WW domain-binding protein 4; WW domain-containing binding protein 4; WW domain-containing-binding protein 4
Gene Aliases: FBP21; FNBP21; WBP4
UniProt ID: (Human) O75554, (Rat) Q5HZF2
Entrez Gene ID: (Human) 11193, (Rat) 114765
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.