Abnova
Manufacturer Code:PAB23372
Catalog # 89-125-070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human WBP4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant WBP4.
For Research Use Only
Protein Aliases: domain-containing binding protein 4; FBP21; formin binding protein 21; Formin-binding protein 21; MGC117310; WBP-4; WW domain-binding protein 4; WW domain-containing binding protein 4; WW domain-containing-binding protein 4
Gene Aliases: FBP21; FNBP21; WBP4
UniProt ID: (Human) O75554
Entrez Gene ID: (Human) 11193
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.