Invitrogen
Manufacturer Code:PA531241
Catalog # PIPA531241
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 290 of Human WBP11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Npw38-binding protein; Npw38-binding protein NpwBP; NpwBP; NPWBP SIPP1 WBP-11 Npw38-binding protein NpwBP SH3 domain-binding protein SNP70 WW domain-binding protein 11 splicing factor that interacts with PQBP-1 and PP1 splicing factor PQBP1 and PP1 interacting; protein phosphatase 1, regulatory subunit 165; SH3 domain-binding protein SNP70; Splicing factor that interacts with PQBP-1 and PP1; splicing factor, PQBP1 and PP1 interacting; WBP-11; WW domain-binding protein 11
Gene Aliases: 2510026P17Rik; D6Wsu113e; NPWBP; PPP1R165; SIPP1; SNP70; WBP-11; WBP11
UniProt ID: (Human) Q9Y2W2, (Mouse) Q923D5, (Rat) Q5PQQ2
Entrez Gene ID: (Human) 51729, (Mouse) 60321, (Rat) 297695
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.