Invitrogen
Manufacturer Code:PA582134
Catalog # PIPA582134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human WASF1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homology of dictyostelium scar 1; KIAA0269 SCAR1 WAVE WAVE1; Protein WAVE-1; Verprolin homology domain-containing protein 1; WAS protein family, member 1; WASP family protein member 1; Wiskott-Aldrich syndrome protein family member 1
Gene Aliases: KIAA0269; SCAR1; WASF1; WAVE; WAVE1
UniProt ID: (Human) Q92558
Entrez Gene ID: (Human) 8936
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.