Invitrogen
Manufacturer Code:PA538348
Catalog # PIPA538348
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic non-phosphopeptide derived from human WAVE1 around the phosphorylation site of Tyr125 (E-T-YP-D-V) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: homology of dictyostelium scar 1; Protein WAVE-1; Verprolin homology domain-containing protein 1; WAS protein family, member 1; WASP family 1; WASP family protein member 1; Wiskott-Aldrich syndrome protein family member 1; Wiskott-Aldrich syndrome verproline-homologous protein 1 Scar1 WASF1
Gene Aliases: AI195380; AI838537; KIAA0269; Scar; SCAR1; WASF1; WAVE; WAVE-1; WAVE1
UniProt ID: (Human) Q92558, (Mouse) Q8R5H6
Entrez Gene ID: (Human) 8936, (Mouse) 83767
Molecular Function: actin family cytoskeletal protein cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.