Invitrogen
Manufacturer Code:PA572617
Catalog # PIPA572617
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 116-144 amino acids from the Central region of human WAS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eczema-thrombocytopenia; thrombocytopenia 1 (X-linked); WAS WASP Wiskott-Aldrich syndrome (eczema-thrombocytopenia) THC IMD2 thrombocytopenia 1 (X-linked) Wiskott-Aldrich syndrome (eczema-thrombocytopenia) protein THC1 Wiskott-Aldrich syndrome protein; WASp; Wiskott-Aldrich syndrome protein; Wiskott-Aldrich syndrome protein homolog
Gene Aliases: IMD2; SCNX; THC; THC1; U42471; WAS; WASP; WASPA
UniProt ID: (Human) P42768, (Mouse) P70315
Entrez Gene ID: (Human) 7454, (Mouse) 22376
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.