Invitrogen
Manufacturer Code:PA537303
Catalog # PIPA537303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length human WASP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eczema-thrombocytopenia; IMD2 SCNX THC THC1 Thrombocytopenia 1 (X linked) U42471 Was WASp WASP Wiskott Aldrich syndrome (eczema thrombocytopenia) Wiskott Aldrichsyndrome protein Wiskott-Aldrich syndrome protein; thrombocytopenia 1 (X-linked); WASp; Wiskott-Aldrich syndrome protein
Gene Aliases: IMD2; SCNX; THC; THC1; WAS; WASP; WASPA
UniProt ID: (Human) P42768
Entrez Gene ID: (Human) 7454
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.