R&D Systems
Manufacturer Code:AF3070
Catalog # AF3070
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human WASP Lys230-Gln310 Accession # P42768 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: eczema-thrombocytopenia; IMD2THC Thrombocytopenia 1 WAS WASP WiskTHC1 wiskthrombocytopenia 1 (X-linked); thrombocytopenia 1 (X-linked); WASp; Wiskott-Aldrich syndrome protein
Gene Aliases: IMD2; SCNX; THC; THC1; WAS; WASP; WASPA
UniProt ID: (Human) P42768
Entrez Gene ID: (Human) 7454
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.