Proteintech
Manufacturer Code:143061AP150UL
Catalog # 50-172-9355
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | WASL Fusion Protein Ag5424 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: N WASP NWASP WASL Wiskott Aldrich syndrome like; N-WASP; Neural Wiskott-Aldrich syndrome protein; Wiskott-Aldrich syndrome-like
Gene Aliases: 2900021I12Rik; 3110031I02Rik; N-WASP; NWASP; TRS4; WASL; WASPB
UniProt ID: (Human) O00401, (Mouse) Q91YD9
Entrez Gene ID: (Human) 8976, (Mouse) 73178, (Rat) 682507
Molecular Function: actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.