Abnova
Manufacturer Code:H00007453A01
Catalog # 89-105-725
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 50 a.a. &bsim 149 a.a) partial recombinant protein with GST tag.,WARS (NP_004175 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant WARS.
For Research Use Only
Protein Aliases: GAMMA-2; hWRS; IFI53; IFP53; Interferon-induced protein 53; T1-TrpRS; T2-TrpRS; TrpRS; tryptophan tRNA ligase 1, cytoplasmic; Tryptophan--tRNA ligase, cytoplasmic; Tryptophanyl-tRNA synthetase
Gene Aliases: GAMMA-2; IFI53; IFP53; WARS; WARS1; WRS
UniProt ID: (Human) P23381
Entrez Gene ID: (Human) 7453
Molecular Function: aminoacyl-tRNA synthetase ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.