Proteintech
Manufacturer Code:163701AP150UL
Catalog # 50-173-0898
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | WAPL WAPAL Fusion Protein Ag9280 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DIF-2; Dioxin-inducible factor 2; FOE Friend of EBNA2 protein KIAA0261 WAPAL WAPL; friend of EBNA2; friend of EBNA2 (Epstein-Barr virus nuclear protein 2); Friend of EBNA2 protein; WAPL cohesin release factor; Wings apart-like protein homolog
Gene Aliases: A530089A20Rik; BC037674; DIF-2; FOE; KIAA0261; WAPAL; WAPL
UniProt ID: (Human) Q7Z5K2, (Mouse) Q65Z40
Entrez Gene ID: (Human) 23063, (Mouse) 218914
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.