R&D Systems
Manufacturer Code:MAB2349
Catalog # MAB2349
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 342603 |
Immunogen | Human plasma-derived Vitronectin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: complement S-protein; complement S-protein epibolin serum spreading factor Serum-spreading factor somatomedin B S-protein V75 vitronectin vitronectin (serum spreading factor somatomedin B complement S-protein) VN VNT VTN; epibolin; S-protein; serum spreading factor; Serum-spreading factor; somatomedin B; Somatomedin-B; V75; Vitronectin; Vitronectin V10 subunit; Vitronectin V65 subunit; VN
Gene Aliases: V75; VN; VNT; VTN
UniProt ID: (Human) P04004
Entrez Gene ID: (Human) 7448
Molecular Function: cell adhesion molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.