R&D Systems
Manufacturer Code:MAB41881100
Catalog # MAB41881100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 884706 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse Vitamin D BP Met1-Ser476 Accession # P21614 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Rat Monoclonal Antibody
Protein Aliases: DBP; DBP DBP/GC DBP-MAF DBPVDBG EC 6.3.1.5 Gc Gc-globulin Gc-MAF GRD3 Group-specific component group-specific component (vitamin D binding protein) hDBP VDB VDBG VDBP vitamin D-binding alpha-globulin vitamin D-binding protein; DBP-maf; epididymis secretory protein Li 51; Gc; Gc protein-derived macrophage activating factor; Gc-globulin; Gc-MAF; Group-specific component; group-specific component (vitamin D binding protein); VDB; vitamin D-binding alpha-globulin; Vitamin D-binding protein; Vitamin D-binding protein-macrophage activating factor
Gene Aliases: DBP; DBP/GC; GC; Gc-MAF; GcMAF; GRD3; HEL-S-51; VDBG; VDBP
UniProt ID: (Human) P02774
Entrez Gene ID: (Human) 2638
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.