Invitrogen
Manufacturer Code:PA532639
Catalog # PIPA532639
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide derived from C-terminus of human vinculin protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMD1W CMH15 MVCL metavinculin; epididymis luminal protein 114; Metavinculin; MV; Vinculin
Gene Aliases: CMD1W; CMH15; HEL114; MV; MVCL; VCL
UniProt ID: (Human) P18206
Entrez Gene ID: (Human) 7414
Molecular Function: actin family cytoskeletal protein cell adhesion molecule cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.