Novus Biologicals
Manufacturer Code:NBP241237
Catalog # NBP241237
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 16 amino acid peptide near the carboxy terminus of human Vinculin. The immunogen is located within amino acids 1060 - 1110 of Vinculin . Amino Acid Squence: LGRTNISDEESEQATE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMD1W CMH15 Metavinculin MVCL vinculin; epididymis luminal protein 114; Metavinculin; MV; Vinculin
Gene Aliases: CMD1W; CMH15; HEL114; MV; MVCL; VCL
UniProt ID: (Human) P18206
Entrez Gene ID: (Human) 7414
Molecular Function: actin family cytoskeletal protein cell adhesion molecule cytoskeletal protein non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.