Novus Biologicals
Manufacturer Code:NBP250081AF350
Catalog # NP25081A350
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | VIL1/1314 |
Immunogen | Recombinant human Villin fragment of 133 amino acid residues (aa179-311) (exact sequence is proprietary) |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: D2S1471 VIL villin 1 villin-1; Villin-1
Gene Aliases: D2S1471; VIL; VIL1
UniProt ID: (Human) P09327
Entrez Gene ID: (Human) 7429
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.