Novus Biologicals
Manufacturer Code:NBP243553
Catalog # NBP243553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 557 |
Immunogen | Recombinant protein encompassing a sequence within the center region of human VAV1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: oncogene vav proto-oncogene vav vav 1 guanine nucleotide exchange factor vav proto-oncogene VAVvav 1 oncogene; Proto-oncogene vav; vav 1 guanine nucleotide exchange factor; vav 1 oncogene
Gene Aliases: VAV; VAV1
UniProt ID: (Human) P15498
Entrez Gene ID: (Human) 7409
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.