R&D Systems
Manufacturer Code:AF3786
Catalog # AF3786
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Vav-1 Trp25-Leu228 Accession # P15498 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: oncogene vav proto-oncogene vav vav 1 guanine nucleotide exchange factor vav proto-oncogene Vav1 vav-T VAVvav 1 oncogene; Proto-oncogene vav; vav 1 guanine nucleotide exchange factor; vav 1 oncogene
Gene Aliases: VAV; VAV1
UniProt ID: (Human) P15498
Entrez Gene ID: (Human) 7409
Molecular Function: G-protein modulator enzyme modulator guanyl-nucleotide exchange factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.